siRNA Details
| Gene Symbol: MAPKBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128608.1 | NP_001122080.1 | 32 | 4659 | ||
| NM_001265611.1 | NP_001252540.1 | 30 | 3810 | |||
| NM_014994.2 | NP_055809.2 | 31 | 4641 | |||
| NR_049761.1 | 30 | 4451 | ||||
| NR_049762.1 | 29 | 4402 | ||||
| XM_006720438.1 | XP_006720501.1 | 30 | 4458 | |||
| XM_006720439.2 | XP_006720502.1 | 18 | 2733 | |||
| XM_011521383.1 | XP_011519685.1 | 31 | 4476 | |||
| XM_017022017.1 | XP_016877506.1 | 30 | 4752 | |||
| GenBank mRNA | AB011168.2 | - | 32 | 4569 | ||
| AB177850.1 | - | 23 | 3483 | |||
| AK302482.1 | - | 29 | 4343 | |||
| AL833267.1 | - | 31 | 4561 | |||
| BC036660.1 | - | 30 | 4456 | 5266613 | ||
| BC113983.1 | - | 30 | 3702 | 40083186 | ||
| BC114493.1 | - | 32 | 4551 | 40083182 | ||
| BX640787.1 | - | 27 | 5018 | |||
| BX648786.1 | - | 31 | 4592 | |||

MAPKBP1
Genomic Map