siRNA Details
| Gene Symbol: FOXL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005250.2 | NP_005241.1 | 1 | 1132 | ||
| GenBank mRNA | BC100027.1 | - | 1 | 1132 | 30920793 | |
| BC117226.1 | - | 1 | 960 | 40125777 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FOXL1| Gene Symbol: FOXL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005250.2 | NP_005241.1 | 1 | 1132 | ||
| GenBank mRNA | BC100027.1 | - | 1 | 1132 | 30920793 | |
| BC117226.1 | - | 1 | 960 | 40125777 | ||