siRNA Details
| Gene Symbol: PDZRN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015009.2 | NP_055824.1 | 3 | 984 | ||
| XM_017005942.1 | XP_016861431.1 | 2 | 979 | |||
| GenBank mRNA | AB029018.1 | - | 3 | 984 | ||
| BC014432.1 | - | 3 | 903 | |||
| BC150623.1 | - | 3 | 971 | 9056993 | ||
| BC152417.1 | - | 3 | 1007 | 8862525 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_018884, HM560981, AK173098, AF127084, AK135776, and others.

PDZRN3
Genomic Map