siRNA Details
| Gene Symbol: PHLPP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289003.1 | NP_001275932.1 | 2 | 798 | ||
| NM_015020.3 | NP_055835.2 | 2 | 451 | |||
| GenBank mRNA | AB023148.2 | - | 2 | 798 | ||
| AK124678.1 | - | 2 | 122 | |||
| BC129927.1 | - | 1 | 78 | 40124720 | ||
| BC129983.1 | - | 1 | 78 | |||
| BX647823.1 | - | 2 | 452 | |||

PHLPP2
Genomic Map