siRNA Details
| Gene Symbol: EMC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271427.1 | NP_001258356.1 | 18 | 2192 | ||
| NM_001271428.1 | NP_001258357.1 | 18 | 2192 | |||
| NM_001271429.1 | NP_001258358.1 | 17 | 2129 | |||
| NM_015047.2 | NP_055862.1 | 18 | 2195 | |||
| XM_005245787.1 | XP_005245844.1 | 19 | 2199 | |||
| XM_005245788.1 | XP_005245845.1 | 19 | 2196 | |||
| GenBank mRNA | AK025853.1 | - | 5 | 556 | ||
| AK075563.1 | - | 17 | 2092 | |||
| AK291618.1 | - | 18 | 2158 | |||
| BC034589.1 | - | 18 | 2188 | 4831005 | ||
| BX648627.1 | - | 18 | 2174 | |||
| BX648708.1 | - | 18 | 2150 | |||
| D42044.2 | - | 18 | 2139 | |||

EMC1
Genomic Map