siRNA Details
| Gene Symbol: EMC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271427.1 | NP_001258356.1 | 2, 3 | 272 | ||
| NM_001271428.1 | NP_001258357.1 | 2, 3 | 272 | |||
| NM_015047.2 | NP_055862.1 | 2, 3 | 272 | |||
| XM_005245787.1 | XP_005245844.1 | 2, 3 | 267 | |||
| XM_005245788.1 | XP_005245845.1 | 2, 3 | 267 | |||
| GenBank mRNA | AK291618.1 | - | 2, 3 | 235 | ||
| BC034589.1 | - | 2, 3 | 265 | 4831005 | ||
| BX648627.1 | - | 2, 3 | 251 | |||
| BX648708.1 | - | 2, 3 | 230 | |||
| D42044.2 | - | 2, 3 | 219 | |||
| DB097686.1 | - | 2, 3 | 272 | |||

EMC1
Genomic Map