siRNA Details
| Gene Symbol: EXPH5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015065.2 | NP_055880.2 | 1 | 137 | ||
| GenBank mRNA | AY099469.1 | - | 1 | 123 | ||
| BC113119.1 | - | 1 | 108 | 40037748 | ||
| BC117701.1 | - | 1 | 108 | 40074153 | ||
| BP358360.1 | - | 1 | 137 | |||

EXPH5
Genomic Map