siRNA Details
| Gene Symbol: NACAD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146334.1 | NP_001139806.1 | 2 | 3125 | ||
| XM_006715674.3 | XP_006715737.1 | 2 | 3674 | |||
| GenBank mRNA | AB002361.1 | - | 1 | 3007 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NACAD| Gene Symbol: NACAD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146334.1 | NP_001139806.1 | 2 | 3125 | ||
| XM_006715674.3 | XP_006715737.1 | 2 | 3674 | |||
| GenBank mRNA | AB002361.1 | - | 1 | 3007 | ||