siRNA Details
| Gene Symbol: TBC1D9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015130.2 | NP_055945.2 | 17, 18 | 3084 | ||
| GenBank mRNA | AB020689.2 | - | 17, 18 | 2887 | ||
| AB449892.1 | - | 17, 18 | 2812 | |||
| AK127020.1 | - | 14, 15 | 2372 | |||
| BC038804.1 | - | 8, 9 | 1198 | |||
| BC146758.1 | - | 17, 18 | 2887 | 9007196 | ||

TBC1D9
Genomic Map