siRNA Details
| Gene Symbol: FLNC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127487.1 | NP_001120959.1 | 41 | 7164 | ||
| NM_001458.4 | NP_001449.3 | 42 | 7263 | |||
| GenBank mRNA | AB208865.1 | - | 19 | 2905 | ||
| AB371585.1 | - | 42 | 7283 | |||
| AF089841.1 | - | 42 | 7103 | |||
| AJ012737.1 | - | 41 | 6973 | |||

FLNC
Genomic Map