siRNA Details
| Gene Symbol: TSPYL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021648.4 | NP_067680.3 | 1 | 785 | ||
| GenBank mRNA | AB018264.1 | - | 1 | 754 | ||
| AK091559.1 | - | 3 | 538 | |||
| AK302542.1 | - | 1 | 788 | |||
| BC009116.1 | - | 1 | 159 | |||
| BC040453.1 | - | 1 | 729 | |||

TSPYL4
Genomic Map