siRNA Details
| Gene Symbol: NUP160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318399.1 | NP_001305328.1 | 3 | 1033 | ||
| NM_015231.2 | NP_056046.1 | 4 | 721 | |||
| NR_134636.1 | 4 | 721 | ||||
| GenBank mRNA | AK026236.1 | - | 3 | 816 | ||
| AK304308.1 | - | 4 | 507 | |||
| AL832029.1 | - | 4 | 511 | |||
| BC009822.2 | - | 3 | 994 | |||
| BC125227.1 | - | 4 | 644 | 40124592 | ||
| BC125228.1 | - | 4 | 644 | 40124593 | ||
| BX647634.1 | - | 4 | 506 | |||
| D83781.1 | - | 4 | 490 | |||

NUP160
Genomic Map