siRNA Details
| Gene Symbol: FMOD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002023.4 | NP_002014.2 | 2 | 1280 | ||
| GenBank mRNA | AK092899.1 | - | 3 | 741 | ||
| AK172740.1 | - | 1 | 979 | |||
| AK291632.1 | - | 2 | 946 | |||
| AK303866.1 | - | 3 | 904 | |||
| BC035281.1 | - | 2 | 957 | 5181410 | ||
| X75546.1 | - | 1 | 896 | |||

FMOD
Genomic Map