siRNA Details
| Gene Symbol: MAN2B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001292038.1 | NP_001278967.1 | 11 | 1456 | ||
| NM_015274.2 | NP_056089.1 | 11 | 1609 | |||
| GenBank mRNA | AB023152.2 | - | 11 | 1568 | ||
| AK025803.1 | - | 3 | 237 | |||
| AK301473.1 | - | 11 | 1441 | |||
| AL832232.1 | - | 11 | 1579 | |||
| AL833071.2 | - | 11 | 1582 | |||
| BC033307.1 | - | 11 | 1578 | 4932398 | ||
| BC094773.1 | - | 11 | 1609 | 30531368 | ||

MAN2B2
Genomic Map