siRNA Details
| Gene Symbol: FAM189A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015307.1 | NP_056122.1 | 3, 4 | 391 | ||
| XM_011521407.2 | XP_011519709.1 | 3, 4 | 320 | |||
| GenBank mRNA | DA188047.1 | - | 3, 4 | 387 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM189A1| Gene Symbol: FAM189A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015307.1 | NP_056122.1 | 3, 4 | 391 | ||
| XM_011521407.2 | XP_011519709.1 | 3, 4 | 320 | |||
| GenBank mRNA | DA188047.1 | - | 3, 4 | 387 | ||