siRNA Details
| Gene Symbol: ICE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015325.2 | NP_056140.1 | 13 | 3871 | ||
| XM_011513999.2 | XP_011512301.1 | 13 | 3871 | |||
| XM_017009285.1 | XP_016864774.1 | 14 | 3960 | |||
| GenBank mRNA | AB023164.3 | - | 13 | 3850 | ||
| AK024400.1 | - | 1 | 801 | |||
| AK095073.1 | - | 1 | 1427 | |||
| AK096615.1 | - | 1 | 2101 | |||
| AL832204.1 | - | 1 | 2342 | |||
| BX538020.1 | - | 1 | 1360 | |||
| CR749441.1 | - | 6 | 3142 | |||

ICE1
Genomic Map