siRNA Details
| Gene Symbol: NCAPH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281710.1 | NP_001268639.1 | 14 | 1832 | ||
| NM_001281711.1 | NP_001268640.1 | 14 | 1793 | |||
| NM_001281712.1 | NP_001268641.1 | 13 | 1612 | |||
| NM_015341.4 | NP_056156.2 | 14 | 1865 | |||
| XM_005263908.3 | XP_005263965.1 | 14 | 1805 | |||
| GenBank mRNA | AK291053.1 | - | 14 | 1806 | ||
| AK299248.1 | - | 14 | 1734 | |||
| AK303725.1 | - | 13 | 1557 | |||
| AK308050.1 | - | 14 | 1773 | |||
| BC024211.2 | - | 14 | 1819 | 3504782 | ||
| D38553.1 | - | 14 | 1795 | |||

NCAPH
Genomic Map