siRNA Details
| Gene Symbol: TPSD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012217.2 | NP_036349.1 | 4 | 637 | ||
| GenBank mRNA | AF206664.1 | - | 3 | 474 | ||
| BC069143.1 | - | 4 | 628 | 7217003 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TPSD1| Gene Symbol: TPSD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012217.2 | NP_036349.1 | 4 | 637 | ||
| GenBank mRNA | AF206664.1 | - | 3 | 474 | ||
| BC069143.1 | - | 4 | 628 | 7217003 | ||