siRNA Details
| Gene Symbol: KCTD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015353.2 | NP_056168.1 | 5 | 807 | ||
| NR_110834.1 | 6 | 688 | ||||
| NR_110835.1 | 7 | 770 | ||||
| GenBank mRNA | AK307083.1 | - | 7 | 725 | ||
| AK310690.1 | - | 2 | 587 | |||
| BC033329.1 | - | 6 | 688 | |||
| D79998.1 | - | 5 | 746 | |||

KCTD2
Genomic Map