siRNA Details
| Gene Symbol: NUP188 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015354.2 | NP_056169.1 | 26 | 2826 | ||
| GenBank mRNA | BC005407.1 | - | 7 | 767 | ||
| BC040352.1 | - | 13 | 1433 | |||
| BC111045.1 | - | 4 | 475 | |||
| BX537774.1 | - | 23 | 2470 | |||
| D79991.1 | - | 26 | 2788 | |||

NUP188
Genomic Map