siRNA Details
| Gene Symbol: SHPK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013276.2 | NP_037408.2 | 7 | 1254 | ||
| GenBank mRNA | AF163573.1 | - | 7 | 1254 | ||
| AK055609.1 | - | 7 | 1180 | |||
| AK057040.1 | - | 1 | 91 | |||
| AK298360.1 | - | 5 | 693 | |||
| AK312428.1 | - | 7 | 1201 | |||
| AL832420.1 | - | 7 | 1193 | |||
| BC020543.1 | - | 7 | 1210 | 4413491 | ||

SHPK
Genomic Map