siRNA Details
| Gene Symbol: ZNF318 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014345.2 | NP_055160.2 | 5 | 2826 | ||
| XM_011514451.2 | XP_011512753.1 | 5 | 2427 | |||
| XM_011514452.2 | XP_011512754.1 | 5 | 3059 | |||
| GenBank mRNA | AF090114.1 | - | 5 | 2829 | ||
| AF121141.1 | - | 5 | 2564 | |||
| AK025505.1 | - | 2 | 1331 | |||
| BC030687.1 | - | 5 | 2861 | |||
| BC098434.1 | - | 5 | 2801 | |||

ZNF318
Genomic Map