siRNA Details
| Gene Symbol: ZNF318 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014345.2 | NP_055160.2 | 10 | 3677 | ||
| XM_011514451.2 | XP_011512753.1 | 10 | 3278 | |||
| GenBank mRNA | AF090114.1 | - | 10 | 3680 | ||
| AF121141.1 | - | 10 | 3415 | |||
| AK025505.1 | - | 7 | 2182 | |||
| BC098434.1 | - | 10 | 3652 | |||

ZNF318
Genomic Map