siRNA Details
| Gene Symbol: MS4A6E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139249.2 | NP_640342.1 | 2 | 337 | ||
| GenBank mRNA | AF333758.1 | - | 2 | 388 | ||
| AF354931.1 | - | 2 | 337 | |||
| AF354932.1 | - | 2 | 221 | |||
| AF354933.1 | - | 2 | 277 | |||
| BC034588.1 | - | 2 | 251 | |||
| BC069322.1 | - | 2 | 337 | 7262144 | ||
| BC101783.1 | - | 2 | 397 | 8069289 | ||

MS4A6E
Genomic Map