siRNA Details
| Gene Symbol: TMEM244 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010876.1 | NP_001010876.1 | 4 | 350 | ||
| XM_011535669.2 | XP_011533971.1 | 5 | 523 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM244| Gene Symbol: TMEM244 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010876.1 | NP_001010876.1 | 4 | 350 | ||
| XM_011535669.2 | XP_011533971.1 | 5 | 523 | |||