siRNA Details
| Gene Symbol: ZNF620 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175888.3 | NP_787084.1 | 3, 4 | 311 | ||
| XM_005265011.3 | XP_005265068.1 | 3, 4 | 3573 | |||
| XM_005265012.3 | XP_005265069.1 | 3, 4 | 3498 | |||
| XM_017006064.1 | XP_016861553.1 | 4, 5 | 3710 | |||
| XM_017006066.1 | XP_016861555.1 | 4, 5 | 3614 | |||
| XM_017006067.1 | XP_016861556.1 | 2, 3 | 209 | |||
| GenBank mRNA | AK131232.1 | - | 3, 4 | 311 | ||
| DA172830.1 | - | 3, 4 | 311 | |||

ZNF620
Genomic Map