siRNA Details
| Gene Symbol: SCML4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198081.4 | NP_932347.2 | 2 | 314 | ||
| XM_011535707.2 | XP_011534009.1 | 2 | 333 | |||
| XM_017010677.1 | XP_016866166.1 | 2 | 381 | |||
| XM_017010678.1 | XP_016866167.1 | 2 | 294 | |||
| XM_017010679.1 | XP_016866168.1 | 2 | 192 | |||
| XM_017010680.1 | XP_016866169.1 | 2 | 260 | |||
| XM_017010681.1 | XP_016866170.1 | 2 | 170 | |||
| XM_017010682.1 | XP_016866171.1 | 2 | 384 | |||
| XM_017010684.1 | XP_016866173.1 | 2 | 386 | |||
| XM_017010685.1 | XP_016866174.1 | 2 | 381 | |||
| XM_017010687.1 | XP_016866176.1 | 2 | 382 | |||
| XM_017010688.1 | XP_016866177.1 | 2 | 390 | |||
| GenBank mRNA | AK303562.1 | - | 2 | 301 | ||
| DC410126.1 | - | 2 | 301 | |||

SCML4
Genomic Map