siRNA Details
| Gene Symbol: TEX40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039496.1 | NP_001034585.1 | 1, 2 | 43 | ||
| GenBank mRNA | AL117564.1 | - | 1, 2 | 46 | ||
| Gene Symbol: KCNK4-TEX40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_133662.1 | 7, 8 | 2030 | |||

TEX40
Genomic Map