siRNA Details
| Gene Symbol: NELFB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015456.4 | NP_056271.3 | 11, 12 | 1698 | ||
| GenBank mRNA | AB033008.1 | - | 3, 4 | 380 | ||
| AF464935.1 | - | 11, 12 | 1503 | |||
| AK022651.1 | - | 9, 10 | 1194 | |||
| AK091056.1 | - | 11, 12 | 1691 | |||
| AL050280.1 | - | 8, 9 | 945 | |||
| BC011892.1 | - | 10, 11 | 1407 | 3941719 | ||
| BC019029.1 | - | 11, 12 | 1657 | |||

NELFB
Genomic Map