siRNA Details
| Gene Symbol: HSPA12A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025015.2 | NP_079291.2 | 9 | 1109 | ||
| XM_005269672.3 | XP_005269729.1 | 10 | 1259 | |||
| XM_005269673.4 | XP_005269730.1 | 10 | 2203 | |||
| XM_011539579.2 | XP_011537881.1 | 11 | 1320 | |||
| XM_011539580.2 | XP_011537882.1 | 10 | 1265 | |||
| XM_017016032.1 | XP_016871521.1 | 7 | 831 | |||
| GenBank mRNA | AB007877.1 | - | 9 | 1109 | ||
| AK294672.1 | - | 8 | 923 | |||
| AK294884.1 | - | 4 | 428 | |||

HSPA12A
Genomic Map