siRNA Details
| Gene Symbol: SGMS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147156.3 | NP_671512.1 | 11 | 2154 | ||
| XM_005269675.1 | XP_005269732.1 | 6 | 1846 | |||
| XM_011539582.2 | XP_011537884.1 | 8 | 1758 | |||
| XM_011539583.2 | XP_011537885.1 | 6 | 1635 | |||
| GenBank mRNA | AB154421.1 | - | 7 | 1537 | ||
| AK026683.1 | - | 7 | 1617 | |||
| AK296233.1 | - | 5 | 821 | |||
| AY280959.1 | - | 11 | 2120 | |||
| AY312431.1 | - | 5 | 1346 | |||
| BC042899.1 | - | 5 | 1415 | 4342258 | ||

SGMS1
Genomic Map