siRNA Details
| Gene Symbol: TMIE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147196.2 | NP_671729.2 | 2 | 308 | ||
| XM_006713097.3 | XP_006713160.1 | 2 | 879 | |||
| GenBank mRNA | AY081842.1 | - | 2 | 308 | ||
| BC126258.1 | - | 2 | 271 | 8991974 | ||
| BC126260.1 | - | 2 | 271 | 8991976 | ||

TMIE
Genomic Map