siRNA Details
| Gene Symbol: TAS2R43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176884.2 | NP_795365.2 | 1 | 350 | ||
| GenBank mRNA | BC117423.1 | - | 1 | 349 | 40125974 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

TAS2R43| Gene Symbol: TAS2R43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176884.2 | NP_795365.2 | 1 | 350 | ||
| GenBank mRNA | BC117423.1 | - | 1 | 349 | 40125974 | |