siRNA Details
| Gene Symbol: CHMP2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001244644.1 | NP_001231573.1 | 5 | 733 | ||
| NM_014043.3 | NP_054762.2 | 6 | 825 | |||
| XM_011533576.2 | XP_011531878.1 | 6 | 656 | |||
| GenBank mRNA | AF151842.1 | - | 6 | 784 | ||
| AK002165.1 | - | 6 | 798 | |||
| AK002180.1 | - | 6 | 803 | |||
| AK222654.1 | - | 6 | 785 | |||
| AK296072.1 | - | 5 | 725 | |||
| AK316585.1 | - | 6 | 789 | |||
| AL080122.1 | - | 6 | 798 | |||
| BC001553.1 | - | 6 | 641 | 3460712 | ||
| CR533456.1 | - | 6 | 565 | |||

CHMP2B
Genomic Map