siRNA Details
| Gene Symbol: UBXN7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015562.1 | NP_056377.1 | 2 | 182 | ||
| XM_011512670.2 | XP_011510972.1 | 2 | 130 | |||
| GenBank mRNA | AB018337.1 | - | 2 | 110 | ||
| AK026451.1 | - | 2 | 135 | |||
| AK129880.1 | - | 2 | 135 | |||
| AK308699.1 | - | 2 | 135 | |||
| BC028986.1 | - | 1 | 14 | |||
| BC047420.1 | - | 2 | 132 | |||
| DA119173.1 | - | 2 | 182 | |||

UBXN7
Genomic Map