siRNA Details
| Gene Symbol: CNTNAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014141.5 | NP_054860.1 | 5 | 1224 | ||
| XM_017011950.1 | XP_016867439.1 | 5 | 1550 | |||
| GenBank mRNA | AB020675.1 | - | 5 | 733 | ||
| AF319045.1 | - | 5 | 848 | |||
| AK315113.1 | - | 5 | 813 | |||
| BC093780.1 | - | 5 | 751 | 7939625 | ||
| BC113373.1 | - | 5 | 751 | 8322425 | ||
| KJ534811.1 | - | 4 | 597 | |||

CNTNAP2
Genomic Map