siRNA Details
| Gene Symbol: DECR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020664.3 | NP_065715.1 | 4 | 403 | ||
| GenBank mRNA | AJ293009.1 | - | 4 | 318 | ||
| AK055839.1 | - | 5 | 1049 | |||
| AK128012.1 | - | 5 | 521 | |||
| AK225961.1 | - | 4 | 572 | |||
| BC010740.1 | - | 4 | 389 | 3877040 | ||
| BC011968.1 | - | 4 | 308 | 3866503 | ||

DECR2
Genomic Map