siRNA Details
| Gene Symbol: ZNF658 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317916.1 | NP_001304845.1 | 4 | 477 | ||
| NM_033160.6 | NP_149350.3 | 4 | 320 | |||
| NR_134255.1 | 3 | 365 | ||||
| XM_005272515.4 | XP_005272572.1 | 4 | 427 | |||
| XM_011545679.2 | XP_011543981.1 | 4 | 251 | |||
| XM_017014614.1 | XP_016870103.1 | 3 | 2613 | |||
| GenBank mRNA | DA120232.1 | - | 4 | 259 | ||

ZNF658
Genomic Map