siRNA Details
| Gene Symbol: LRRC32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128922.1 | NP_001122394.1 | 3 | 2105 | ||
| NM_005512.2 | NP_005503.1 | 3 | 2210 | |||
| XM_005273902.3 | XP_005273959.1 | 3 | 2299 | |||
| XM_005273903.4 | XP_005273960.2 | 3 | 2192 | |||
| XM_011544935.2 | XP_011543237.1 | 3 | 2501 | |||
| XM_017017527.1 | XP_016873016.1 | 3 | 2705 | |||
| GenBank mRNA | AK314971.1 | - | 4 | 2142 | ||
| BC052210.1 | - | 3 | 2102 | 4825609 | ||
| BC070079.1 | - | 3 | 2105 | 30344529 | ||
| Z24680.1 | - | 3 | 2061 | |||

LRRC32
Genomic Map