siRNA Details
| Gene Symbol: RGS22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286692.1 | NP_001273621.1 | 7 | 856 | ||
| NM_001286693.1 | NP_001273622.1 | 5 | 751 | |||
| NM_015668.4 | NP_056483.3 | 7 | 892 | |||
| XM_005250856.2 | XP_005250913.1 | 7 | 738 | |||
| XM_005250857.2 | XP_005250914.1 | 7 | 738 | |||
| XM_005250858.2 | XP_005250915.1 | 7 | 738 | |||
| XM_005250860.2 | XP_005250917.1 | 7 | 738 | |||
| XM_006716540.2 | XP_006716603.1 | 7 | 1006 | |||
| XM_011516957.1 | XP_011515259.1 | 8 | 1187 | |||
| XM_011516958.1 | XP_011515260.1 | 7 | 1085 | |||
| XM_011516959.2 | XP_011515261.1 | 7 | 738 | |||
| XM_017013309.1 | XP_016868798.1 | 7 | 738 | |||
| XM_017013310.1 | XP_016868799.1 | 7 | 738 | |||
| XM_017013311.1 | XP_016868800.1 | 1 | 63 | |||
| GenBank mRNA | AK292559.1 | - | 7 | 702 | ||
| AK302475.1 | - | 4 | 512 | |||
| AK309194.1 | - | 4 | 499 | |||
| AY009106.1 | - | 7 | 685 | |||
| BC036665.1 | - | 7 | 743 | 5268331 | ||
| BC047060.1 | - | 7 | 776 | |||
| BC092411.1 | - | 7 | 889 | 30386231 | ||

RGS22
Genomic Map