siRNA Details
| Gene Symbol: TIPRL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031800.2 | NP_001026970.1 | 1 | 275 | ||
| NM_152902.4 | NP_690866.1 | 1 | 275 | |||
| GenBank mRNA | AB097034.1 | - | 1 | 158 | ||
| AK313520.1 | - | 1 | 275 | |||
| AL049670.1 | - | 1 | 187 | |||
| BC009506.2 | - | 1 | 65 | 3532344 | ||

TIPRL
Genomic Map