siRNA Details
Gene Symbol: TMEM251 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098621.1 | NP_001092091.1 | 2 | 649 | ||
NM_015676.1 | NP_056491.1 | 2 | 275 | |||
GenBank mRNA | AL080118.1 | - | 2 | 244 | ||
BC030119.2 | - | 2 | 242 | |||
CX785861.1 | - | 2 | 649 |
