siRNA Details
| Gene Symbol: EHF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001206615.1 | NP_001193544.1 | 3, 4 | 529 | ||
| NM_001206616.1 | NP_001193545.1 | 3, 4 | 530 | |||
| NM_012153.5 | NP_036285.2 | 3, 4 | 529 | |||
| XM_005252857.1 | XP_005252914.1 | 3, 4 | 530 | |||
| XM_005252860.1 | XP_005252917.1 | 3, 4 | 383 | |||
| XM_005252861.1 | XP_005252918.1 | 3, 4 | 530 | |||
| XM_005252862.1 | XP_005252919.1 | 3, 4 | 399 | |||
| XM_011519983.1 | XP_011518285.1 | 3, 4 | 555 | |||
| XM_011519984.1 | XP_011518286.1 | 4, 5 | 635 | |||
| XM_011519985.1 | XP_011518287.1 | 3, 4 | 420 | |||
| XM_017017534.1 | XP_016873023.1 | 3, 4 | 530 | |||
| GenBank mRNA | AF124438.1 | - | 3, 4 | 435 | ||
| AF124439.1 | - | 3, 4 | 435 | |||
| AF170583.1 | - | 3, 4 | 439 | |||
| AF203977.1 | - | 3, 4 | 450 | |||
| AF212848.1 | - | 3, 4 | 469 | |||
| AK309318.1 | - | 3, 4 | 555 | |||
| AK310867.1 | - | 3, 4 | 530 | |||
| AL703839.1 | - | 3, 4 | 502 | |||
| AY882601.1 | - | 2, 3 | 338 | |||
| BC038995.2 | - | 3, 4 | 489 | 6055934 | ||
| DA460976.1 | - | 3, 4 | 528 | |||
| DB214606.1 | - | 4, 5 | 420 | |||

EHF
Genomic Map