siRNA Details
| Gene Symbol: ITGB1BP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303277.1 | NP_001290206.1 | 2 | 173 | ||
| NM_012278.2 | NP_036410.1 | 5 | 426 | |||
| XM_017029405.1 | XP_016884894.1 | 3 | 696 | |||
| GenBank mRNA | AF110225.1 | - | 5 | 380 | ||
| AF140690.1 | - | 5 | 396 | |||
| BC108901.2 | - | 4 | 653 | 40004692 | ||
| BP372847.1 | - | 2 | 171 | |||
| DA895605.1 | - | 5 | 429 | |||

ITGB1BP2
Genomic Map