siRNA Details
| Gene Symbol: RGL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153615.1 | NP_705843.1 | 10 | 2504 | ||
| XM_017028767.1 | XP_016884256.1 | 4 | 448 | |||
| GenBank mRNA | AK131340.1 | - | 13 | 3109 | ||
| AY101396.1 | - | 10 | 2504 | |||
| BC047947.1 | - | 2 | 1442 | |||
| BC101108.3 | - | 9 | 2184 | 40011557 | ||
| BC101109.2 | - | 9 | 2184 | 40011564 | ||
| BC101110.3 | - | 10 | 2059 | |||
| HY101412.1 | - | 5 | 345 | |||

RGL4
Genomic Map