siRNA Details
| Gene Symbol: PGBD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017434.1 | NP_001017434.1 | 3 | 345 | ||
| NM_170725.2 | NP_733843.1 | 3 | 1022 | |||
| XM_005270333.1 | XP_005270390.1 | 4 | 1121 | |||
| XM_011544159.2 | XP_011542461.1 | 3 | 1023 | |||
| XM_011544161.2 | XP_011542463.1 | 3 | 1963 | |||
| GenBank mRNA | AK123219.1 | - | 3 | 392 | ||
| AK298248.1 | - | 4 | 1122 | |||
| BC063785.1 | - | 3 | 1027 | 6173868 | ||
| BX647065.1 | - | 3 | 334 | |||

PGBD2
Genomic Map