siRNA Details
| Gene Symbol: RNF11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014372.4 | NP_055187.1 | 3 | 793 | ||
| GenBank mRNA | AB024703.1 | - | 3 | 436 | ||
| AF151881.1 | - | 3 | 436 | |||
| AK293047.1 | - | 5 | 622 | |||
| AK313140.1 | - | 3 | 555 | |||
| BC020964.2 | - | 3 | 388 | 3917224 | ||
| BC047654.1 | - | 3 | 761 | 5261966 | ||

RNF11
Genomic Map