siRNA Details
| Gene Symbol: SND1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014390.2 | NP_055205.2 | 5 | 755 | ||
| XM_017011987.1 | XP_016867476.1 | 5 | 784 | |||
| GenBank mRNA | AB209510.1 | - | 5 | 693 | ||
| AK096583.1 | - | 6 | 680 | |||
| AK304181.1 | - | 5 | 647 | |||
| AK312313.1 | - | 5 | 709 | |||
| BC017180.2 | - | 5 | 711 | 3345037 | ||
| BT009785.1 | - | 5 | 529 | |||
| U22055.1 | - | 5 | 721 | |||

SND1
Genomic Map