siRNA Details
Gene Symbol: FAM184B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015688.1 | NP_056503.1 | 5 | 1403 | ||
GenBank mRNA | AB033102.1 | - | 5 | 1216 | ||
AK308144.1 | - | 5 | 1403 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FAM184B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015688.1 | NP_056503.1 | 5 | 1403 | ||
GenBank mRNA | AB033102.1 | - | 5 | 1216 | ||
AK308144.1 | - | 5 | 1403 |