siRNA Details
| Gene Symbol: SIT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014450.2 | NP_055265.1 | 3 | 370 | ||
| GenBank mRNA | AA310964.1 | - | 3 | 371 | ||
| AJ010059.1 | - | 3 | 360 | |||
| AK314758.1 | - | 3 | 351 | |||
| BC102029.1 | - | 3 | 368 | 40031396 | ||
| BC104491.1 | - | 3 | 368 | 40031403 | ||
| BC107484.1 | - | 3 | 368 | 40031398 | ||
| BM919490.1 | - | 3 | 210 | |||
| JQ924063.1 | - | 2 | 444 | |||

SIT1
Genomic Map